Skip to Content
Merck
All Photos(1)

Key Documents

V0255

Sigma-Aldrich

[pGlu4, (Cys2)6, Arg8]-Vasopressin Fragment 4-9 acetate salt

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H46N12O10S2 · xC2H4O2
CAS Number:
Molecular Weight:
774.87 (free base basis)
MDL number:
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(O)=O.N[C@@H](CSSC[C@H](NC(=O)[C@H](CC(N)=O)NC(=O)[C@@H]1CCC(=O)N1)C(=O)N2CCC[C@H]2C(=O)N[C@@H](CCCNC(N)=N)C(=O)NCC(N)=O)C(O)=O

Gene Information

human ... AVP(551)

Amino Acid Sequence

pGlu-Asn-Cys(Cys)-Pro-Arg-Gly-NH2

Biochem/physiol Actions

A more potent neuropeptide than AVP with selective effects on memory and related processes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Burbach et al.
Science (New York, N.Y.), 221(4617), 1310-1312 (1983-09-23)
A peptide that accumulated as the major product during the proteolysis of arginine vasopressin by rat brain synaptic membranes was isolated and its structure was shown to be the hexapeptide pGlu-Asn-Cys(Cys)-Pro-Arg-Gly-NH2. When administered intracerebroventricularly in extremely low doses, this vasopressin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service