Skip to Content
Merck
All Photos(1)

Documents

SRP6456

Sigma-Aldrich

EPHB4 human

recombinant, expressed in HEK 293 cells, ≥97% (SDS-PAGE)

Synonym(s):

Ephrin-type-B-receptor-4, HTK, MYK1, TYRO11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥97% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 57.8 kDa
observed mol wt 65-70 kDa by SDS-PAGE (DTT- reduced.)

packaging

pkg of 20 and 100 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... EPHB4(2050)

General description

Ephrin type-B receptor 4 (EPHB4) is also known as HTK, MYK1 and TYRO11,is a member of Eph family. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The protein encoded by EPHB4 binds to Ephrin-B2 and plays an essential role in vascular development. EPHB4 and its ligand ephrin-B2 are specifically expressed on venous and arterial endothelial cells, respectively, and play an essential role in vascular development via bidirectional signals. The forward EPHB4 signaling inhibits cell adhesion, chemotaxis, angiogenesis and tumor growth. In contrast, the reverse Ephrin-B2 signaling exerts the opposite effect. It has been reported that aberrant expression of EPHB4 is associated with prostate cancer and highly malignant breast cancers, accordingly, EPHB4 has potential application as a therapeutic candidate.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service