Skip to Content
Merck
All Photos(1)

Key Documents

SRP6171

Sigma-Aldrich

IL-3 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

HCGF, MGC79398, Mast cell growth factor (MCGF), Multi-CSF, P-cell stimulation factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

17-45 kDa (monomer, glycosylated)

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... IL3(3562)

General description

IL3 is a potent growth promoting cytokine. This cytokine is capable of supporting the proliferation of a broad range of hematopoietic cell types. It stimulates colony formation of megakaryocytes, neutrophils, and macrophages from bone marrow cultures. It is also involved in a variety of cell activities such as cell growth, differentiation and apoptosis. This cytokine has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders.

Physical form

Lyophilized from a PBS solution.

Preparation Note

Centrifuge the vial prior to opening.

Reconstitution

Reconstitute in sterile PBS containing 0.1% endotoxin-free recombinant human serum albumin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Yousefi et al.
Allergy, 70(9), 1184-1188 (2015-06-05)
Basophils are primarily associated with immunomodulatory functions in allergic diseases and parasitic infections. Recently, it has been demonstrated that both activated human and mouse basophils can form extracellular DNA traps (BETs) containing mitochondrial DNA and granule proteins. In this report
Hai Zhou et al.
Thrombosis and haemostasis, 113(5), 1021-1034 (2015-01-09)
Impaired megakaryocyte maturation and insufficient platelet production have been shown to participate in the pathogenesis of immune thrombocytopenia (ITP). Our previous study demonstrated that low expression of tumour necrosis factor-related apoptosis-inducing ligand (TRAIL) in megakaryocytes contributed to impaired platelet production
Amy H Lin et al.
Journal of virology, 88(17), 10066-10077 (2014-06-27)
We developed a Moloney mouse leukemia virus (MLV)-based retroviral replicating vector (RRV), Toca 511, which has displayed tumor specificity in resected brain tumor material and blood in clinical trials. Here, we investigated the interaction between Toca 511 and human host
Kerstin Kapp et al.
Molecular therapy. Nucleic acids, 3, e170-e170 (2014-06-25)
Toll-like receptors are sensing modulators of the innate immune system. One member of this protein family, Toll-like receptor (TLR)-9, is increasingly being investigated as therapeutic target for infectious diseases and cancer. Double-Stem Loop ImmunoModulator (dSLIM) is a new TLR-9 agonist
Orit Uziel et al.
Biochemical and biophysical research communications, 450(1), 274-282 (2014-06-08)
Treatment with erythropoietin (EPO) in several cancers is associated with decreased survival due to cancer progression. Due to the major importance of telomerase in cancer biology we hypothesized that some of these effects may be mediated through EPO effect on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service