Skip to Content
Merck
All Photos(1)

Documents

SML1805

Sigma-Aldrich

APS-2-79 hydrochloride

≥98% (HPLC)

Synonym(s):

6,7-Dimethoxy-N-(2-methyl-4-phenoxyphenyl)quinazolin-4-amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H21N3O3 · HCl
CAS Number:
Molecular Weight:
423.89
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=CC(OC2=CC=CC=C2)=CC=C1NC3=NC=NC4=CC(OC)=C(OC)C=C43.[H]Cl

Biochem/physiol Actions

APS-2-79 is a stabilizer of the Kinase suppressor of Ras (KSR) inactive state, resulting in inhibition of oncogenic Ras signaling, antagonizing the Ras–MAPK pathway. KSR is a MAPK scaffold that is regulated allosterically by dimerization with RAF. APS-2-79 was found to bind to the KSR active site with an IC50 value of 120 nM, antagonizing MEK (MAPKK) phosphorylation by RAF. In Ras mutant cell lines, APS-2-79 increased the potency of several MEK inhibitors..

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service