Skip to Content
Merck
All Photos(1)

Key Documents

SML0300

Sigma-Aldrich

Aib-1 trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

Tyr-Aib-Trp-Phe trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H37N5O6 · xC2HF3O2
CAS Number:
Molecular Weight:
599.68 (free base basis)
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

lyophilized solid

color

white to beige

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC(C)(NC(=O)[C@@H](N)Cc1ccc(O)cc1)C(=O)N[C@@H](Cc2cc3ccccc3[nH]2)C(=O)N[C@@H](Cc4ccccc4)C(O)=O

InChI

1S/C33H37N5O6.C2HF3O2/c1-33(2,38-29(40)25(34)16-21-12-14-24(39)15-13-21)32(44)37-27(19-23-18-22-10-6-7-11-26(22)35-23)30(41)36-28(31(42)43)17-20-8-4-3-5-9-20;3-2(4,5)1(6)7/h3-15,18,25,27-28,35,39H,16-17,19,34H2,1-2H3,(H,36,41)(H,37,44)(H,38,40)(H,42,43);(H,6,7)/t25-,27-,28-;/m0./s1

InChI key

NELSAMPJZPTPEG-MDQBHWJISA-N

Biochem/physiol Actions

Aib-1 is an inhibitor of β-Amyloid oligomerization.
Aib-1 is an inhibitor of β-Amyloid oligomerization. Aib-1 is an endomorphin analog more stable to enzymatic degradation and incorporating the β-breaker 2 aminoisobutyric acid. It interacted with Aβ and markedly inhibited the formation of toxic oligomer and fibril growth using in vitro assays, prevented Aβ toxicity in neuronal PC12 cells, and increased longevity in a fly model of Alzheimer′s disease. Aib-1 activity is thought to be due to multi-mode interactions including aromatic, hydrophobic, and polar contacts with Aβ.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service