Skip to Content
Merck
All Photos(2)

Documents

SAB5702346

Sigma-Aldrich

Anti-Complement C4A Antibody, clone 4C3F2, Rabbit Monoclonal

Synonym(s):

C4, C4A2, C4A3, C4A4, C4A6, C4AD, C4S, CO4, CPAMD2, RG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

4C3F2, monoclonal

form

liquid

mol wt

200 kDa

species reactivity

human

concentration

0.5 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

HREELVYELNPLDHRGRTLEIPGNSDPNMIPDGDFNSYVRVTASDPLDTLGSEGALSPGGVASLLRLPRGCGEQTMIYLAPTLAASRYLDKTEQWSTLPPETKDHAVDLIQKGYMRIQQFRKADGSYAAWLSRDSSTWLTAFVLKVLSLAQEQVGGSPEKLQETSNWLLSQQQADGSFQDPCPVLDRSMQGGLVGNDETVALTAFVTIALHHGLAVFQDEGAEPLKQRVEASISKANSFLGEKASAGLLGAHAAAITAYALTLTKAPVDLLGVAHNNLMAMAQETGDNLYWGSVTGSQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... C4A(720)

General description

This gene encodes the acidic form of complement factor 4, part of the classical activation pathway. The protein is expressed as a single chain precursor which is proteolytically cleaved into a trimer of alpha, beta, and gamma chains prior to secretion. The trimer provides a surface for interaction between the antigen-antibody complex and other complement components. The alpha chain is cleaved to release C4 anaphylatoxin, an antimicrobial peptide and a mediator of local inflammation. Deficiency of this protein is associated with systemic lupus erythematosus and type I diabetes mellitus. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6. Varying haplotypes of this gene cluster exist, such that individuals may have 1, 2, or 3 copies of this gene. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Nov 2014]

Immunogen

PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Recombinant fusion protein containing a sequence corresponding to amino acids 940-1237 of human Complement C4A (P0C0L4).

Application

WB, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service