Skip to Content
Merck
All Photos(1)

Key Documents

SAB5300324

Sigma-Aldrich

Monoclonal Anti-CHD3 antibody produced in mouse

clone 2G4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

CHD3, Mi-2a, Mi2-ALPHA, ZFH

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
₩701,162

₩701,162


Estimated to ship onApril 25, 2025Details



Select a Size

Change View
100 μG
₩701,162

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

₩701,162


Estimated to ship onApril 25, 2025Details


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2G4, monoclonal

form

buffered aqueous solution

mol wt

226 kDa

species reactivity

human, mouse

technique(s)

direct ELISA: 1:10,000
flow cytometry: 1:200-1:400
immunohistochemistry: 1:200-1:1,000
indirect immunofluorescence: 1:200-1:1,000
western blot: 1:500-1:2,000

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHD3(1107)

Related Categories

Immunogen

Purified recombinant fragment of human CHD3 expressed in E.coli.
Mouse monoclonal antibody raised against CHD3

Physical form

Purified antibody in PBS with 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pasano Bojang et al.
Molecular oncology, 12(8), 1342-1357 (2018-05-31)
Long interspersed nuclear element-1 (LINE-1 or L1) reactivation is linked to poor prognosis in non-small-cell lung carcinoma (NSCLC), but the molecular bases of this response remain largely unknown. In this report, we show that challenge of human bronchial epithelial cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service