Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500059

Sigma-Aldrich

Anti-Swine H1N1 Hemagglutinin antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Swine flu H1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
₩639,552

₩639,552


Please contact Customer Service for Availability
New, lower price on this item!


Select a Size

Change View
100 μG
₩639,552

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

₩639,552


Please contact Customer Service for Availability
New, lower price on this item!

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

viral

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Immunogen

Hemagglutinin antibody was raised against a synthetic peptide from the novel swine influenza Hemagglutinin protein. The peptide sequence is unique from the peptide sequence for product SAB3500061 and SAB3500063. This antibody is a cognate pair with product SAB3500058.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA. In early 2009, a novel swine-origin influenza A (H1N1) virus was identified in specimens obtained from patients in Mexico and the United States. The virus spread quickly around the world and on June 11, 2009, the World Health Organization declared it a pandemic. Influenza A virus has one of sixteen possible Hemagglutinin (HA) surface proteins and one of nine possible Neuraminidase (NA) surface proteins. The Hemagglutinin protein facilitates viral attachment while Neuraminidase is involved in viral release. These proteins also elicit immune responses that prevent infection or independently reduce viral replication. The genetic make-up of this swine flu virus is unlike any other: it is an H1N1 strain that combines a triple assortment first identified in 1998 including human, swine, and avian influenza with two new pig H3N2 virus genes from Eurasia, themselves of recent human origin. The distinct antigenic properties of the new swine virus compared with seasonal influenza A (H1N1) virus suggest that human immunity against new swine influenza virus is limited, although the age distribution of reported cases suggests some degree of protection in older age groups. This antibody is specific for the novel swine influenza Hemagglutinin and will not recognize the corresponding Hemagglutinin sequence from the seasonal H1N1 influenza (A/Brisbane/59/2007 (H1N1)).

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500059.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anastasia Shevtsova-Horoz et al.
Virus research, 199, 31-41 (2015-01-24)
The Paramyxovirus membrane associated proteins are composed of two integral membrane glycoproteins, HN (H, G) and F, and of a matrix protein (M) carpeting the membrane inner layer. For Sendai virus (SeV), F and M have been proposed to form
Naif Khalaf Alharbi et al.
PloS one, 10(5), e0127978-e0127978 (2015-05-27)
Recombinant modified vaccinia virus Ankara (MVA) has been used to deliver vaccine candidate antigens against infectious diseases and cancer. MVA is a potent viral vector for inducing high magnitudes of antigen-specific CD8+ T cells; however the cellular immune responses to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service