Skip to Content
Merck
All Photos(1)

Key Documents

SAB1412023

Sigma-Aldrich

ANTI-EMX2 antibody produced in mouse

clone 2A10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

EMX2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
₩618,643

₩618,643


Estimated to ship onApril 16, 2025Details



Select a Size

Change View
100 μG
₩618,643

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

₩618,643


Estimated to ship onApril 16, 2025Details


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2A10, monoclonal

form

buffered aqueous solution

mol wt

antigen 36.52 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EMX2(2018)

General description

This gene encodes a homeobox-containing transcription factor that is the homolog to the ′empty spiracles′ gene in Drosophila. Research on this gene in humans has focused on its expression in three tissues: dorsal telencephalon, olfactory neuroepithelium, and urogenetial system. It is expressed in the dorsal telencephalon during development in a low rostral-lateral to high caudal-medial gradient and is proposed to pattern the neocortex into defined functional areas. It is also expressed in embryonic and adult olfactory neuroepithelia where it complexes with eukaryotic translation initiation factor 4E (eIF4E) and possibly regulates mRNA transport or translation. In the developing urogenital system, it is expressed in epithelial tissues and is negatively regulated by HOXA10. Alternative splicing results in multiple transcript variants encoding distinct proteins

Immunogen

EMX2 (NP_004089, 103 a.a. ~ 200 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
HSPHPLFASQQRDPSTFYPWLIHRYRYLGHRFQGNDTSPESFLLHNALARKPKRIRTAFSPSQLLRLEHAFEKNHYVVGAERKQLAHSLSLTETQVKV

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service