Skip to Content
Merck
All Photos(1)

Key Documents

SAB1411648

Sigma-Aldrich

Anti-FBXO44 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp781J0852, FBG3, FBX30, FBX6A, Fbx44, Fbxo6a, MGC14140

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
₩618,643

₩618,643


Estimated to ship onApril 11, 2025Details



Select a Size

Change View
100 μG
₩618,643

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

₩618,643


Estimated to ship onApril 11, 2025Details


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 25.7 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FBXO44(93611)

General description

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. It is also a member of the NFB42 (neural F Box 42 kDa) family, similar to F-box only protein 2 and F-box only protein 6. Four alternatively spliced transcript variants encoding two distinct isoforms have been found for this gene. (provided by RefSeq)

Immunogen

FBXO44 (NP_904319.1, 1 a.a. ~ 224 a.a) full-length human protein.

Sequence
MAVGNINELPENILLELFTHVPARQLLLNCRLVCSLWRDLIDLVTLWKRKCLREGFITEDWDQPVADWKIFYFLRSLHRNLLHNPCAEEGFEFWSLDVNGGDEWKVEDLSRDQRKEFPNDQVRSQARLRVQVPAVRSAPVVRARASGDLPARPGDHPAEERCQVEGGLPHILQLPARRPLHLVSARRRGHSLLGRLVRPEGHQQQHHHRAPAALTPPEPPSAEP

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service