Skip to Content
Merck
All Photos(1)

Key Documents

P9204

Sigma-Aldrich

N6-Phenoxyacetyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyadenosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H37N5O7
CAS Number:
Molecular Weight:
687.74
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

97% (TLC)

form

solid

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(OC2CC(OC2COC(=O)COc3ccccc3)n4cnc5c(N)ncnc45)(c6ccccc6)c7ccc(OC)cc7

InChI

1S/C39H37N5O7/c1-46-29-17-13-27(14-18-29)39(26-9-5-3-6-10-26,28-15-19-30(47-2)20-16-28)51-32-21-34(44-25-43-36-37(40)41-24-42-38(36)44)50-33(32)22-49-35(45)23-48-31-11-7-4-8-12-31/h3-20,24-25,32-34H,21-23H2,1-2H3,(H2,40,41,42)

InChI key

NLUGRLLZQAIJEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service