Skip to Content
Merck
All Photos(1)

Documents

OH30251541GB

Ohaus® MB27 Mositure Analyzer

Model MB27, weighing capacity 90g, resolution: 0.001 g, AC/DC input 220/240 V, UK plug

Synonym(s):

Ohaus® Moisture Analyzer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111500
NACRES:
NB.13

feature

CE compliant

packaging

pack of 1 ea

manufacturer/tradename

Ohaus® 30251541GB

weighing capacity measuring range

90g, resolution: 0.001 g

Looking for similar products? Visit Product Comparison Guide

General description

Combining Speed and Precision in One Moisture Analyzer

Application

Percent Moisture Determination, Percent Solid Determination, Weighing

Features and Benefits

Colour touchscreen display with intuitive icon-based navigation Fast halogen heating technology provides uniform drying for precise and repeatable results Analysis made simple: tare, add your sample, press start and see your results Robust hou

Legal Information

Ohaus is a registered trademark of Ohaus Corp.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service