Skip to Content
Merck
All Photos(1)

Key Documents

N7909

Sigma-Aldrich

1-(2-Nitrophenyl)ethyl phosphate diammonium salt

Synonym(s):

1-(2-Nitrophenyl)ethyl phosphate, diammonium salt, NPE-caged phosphate diammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH(CH3)OP(O)(O-)2 · 2NH3+
Molecular Weight:
317.28
MDL number:
UNSPSC Code:
12171500

SMILES string

[NH4+].[NH4+].CC(OP([O-])([O-])=O)c1ccccc1[N+]([O-])=O

General description

Cell permeable source of inorganic phosphate. UV light photolysis (320-342nm) releases intracellular Pi for transient kinetic studies of phosphorylation.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service