N0262
2-Amino-1,5-naphthalenedisulfonic acid sodium salt
Synonym(s):
2-Naphthylamine-1,5-disulfonic acid sodium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H9NO6S2 · xNa+
CAS Number:
Molecular Weight:
303.31 (free acid basis)
Beilstein:
2888645
EC Number:
MDL number:
UNSPSC Code:
12352200
Recommended Products
storage temp.
2-8°C
SMILES string
Nc1ccc2c(cccc2c1S(O)(=O)=O)S(O)(=O)=O
Application
Reported to be useful in sequential analysis of non-polar peptides.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A new, improved technique for automated sequencing of non-polar peptides.
J A Foster et al.
Biochemical and biophysical research communications, 53(1), 70-74 (1973-07-02)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service