Skip to Content
Merck
All Photos(1)

Documents

MTCP80255S

MTC Bio Reservoir

capacity 25 mL, non-sterile, pack of 200 ea (40 x bags 5 ea)

Synonym(s):

cryo tube storage box, reagent dish, reagent holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121522
NACRES:
NB.12

sterility

non-sterile

packaging

pack of 200 ea (40 x bags 5 ea)

manufacturer/tradename

MTC Bio P8025-5S

capacity

25 mL

Looking for similar products? Visit Product Comparison Guide

General description

Reservoir, 25ml sterile, 5/ sterile bag, 200/cs

Features and Benefits

  • Graduated inner walls
  • Micro-trough U-shaped bottoms
  • Pouring spouts on all 4 corners

Legal Information

MTC is a trademark of MTC Bio

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service