Skip to Content
Merck
All Photos(1)

Documents

MAK439

Sigma-Aldrich

Glycolysis Assay Kit

Sufficient for 100 colorimetric tests

Synonym(s):

Cellular Glycolysis Detection Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

detection method

colorimetric

relevant disease(s)

diabetes; cancer; endocrinological disorders

storage temp.

−20°C

Related Categories

General description

Glycolysis is one of the major metabolic pathways cells undergo to produce energy and results in the production of pyruvate. One of the eventual end products of this process is lactate dehydrogenase-mediated conversion of pyruvate to L-lactate via lactic acid fermentation. This enzymatic mechanism allows L-lactate to serve as an indicator of glycolysis.

Application

The Glycolysis Assay Kit may be used for:
  • Cancer Research
  • Diabetes Research
  • Cardiovascular Disease Research

Features and Benefits

Broad Detection Range: Detect L-lactate levels accurately up to 10 mM, using only 5 μL of sample in a 96-well plate setup, ensuring comprehensive analysis with minimal sample requirement.

Simplified Process: Experience a streamlined process with the addition of only a single working reagent and a room temperature reaction, reducing complexity and saving valuable time and effort.

Compatibility with High-Throughput Systems: Easily incorporate our kit into high-throughput handling systems, ensuring smooth and accurate processing, enhancing efficiency in your laboratory workflow.

Suitability

The kit is suitable for the direct determination of L-Lactate produced by glycolysis in cell media samples and for the screening of glycolysis inhibitors.

Principle

The Glycolysis Assay Kit is based on measuring the production of L-Lactate from glycolysis in cells. L-Lactate that is secreted into the cell media is quantified using a coupled reaction involving the lactate dehydrogenase catalyzed oxidation of L-lactate that generates pyruvate and NADH, which subsequently reduces a formazan dye. The intensity of the reduced dye, measured at 565 nm, is directly proportional to the L-lactate concentration in the sample, which in turn is directly proportional to the glycolytic rate of the cells.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service