Skip to Content
Merck
All Photos(1)

Key Documents

M6913

Sigma-Aldrich

Myelin Basic Protein Fragment 4-14

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H103N21O17
CAS Number:
Molecular Weight:
1390.59
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

mouse

Assay

≥95% (HPLC)

form

solid

composition

Peptide content, ≥65%

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C(Cc1ccc(O)cc1)NC(=O)C(CCCCN)NC(=O)C(CO)NC(=O)C(CCCNC(N)=N)NC(=O)C(CCC(N)=O)NC(=O)C(CO)NC(=O)C2CCCN2C(=O)C(CCCNC(N)=N)NC(=O)C(CCCCN)NC(=O)C(N)CCC(N)=O)C(O)=O

InChI

1S/C60H103N21O17/c1-32(2)28-42(58(97)98)78-53(92)41(29-33-15-17-34(84)18-16-33)77-50(89)37(11-4-6-24-62)74-54(93)43(30-82)79-51(90)38(12-7-25-70-59(66)67)73-52(91)39(20-22-47(65)86)75-55(94)44(31-83)80-56(95)45-14-9-27-81(45)57(96)40(13-8-26-71-60(68)69)76-49(88)36(10-3-5-23-61)72-48(87)35(63)19-21-46(64)85/h15-18,32,35-45,82-84H,3-14,19-31,61-63H2,1-2H3,(H2,64,85)(H2,65,86)(H,72,87)(H,73,91)(H,74,93)(H,75,94)(H,76,88)(H,77,89)(H,78,92)(H,79,90)(H,80,95)(H,97,98)(H4,66,67,70)(H4,68,69,71)

InChI key

KISWVXRQTGLFGD-UHFFFAOYSA-N

Gene Information

mouse ... Mbp(17196)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Gln-Lys-Arg-Pro-Ser-Gln-Arg-Ser-Lys-Tyr-Leu

Substrates

Protein kinase C substrate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Yasuda et al.
Biochemical and biophysical research communications, 166(3), 1220-1227 (1990-02-14)
Among various phosphate acceptor proteins and peptides so far tested, a synthetic peptide having the sequence surrounding Ser(8) of myelin basic protein, Gln-Lys-Arg-Pro-Ser(8)-Gln-Arg-Ser-Lys-Tyr-Leu, (MBP4-14), is the most specific and convenient substrate which can be used for selective assay of protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service