Skip to Content
Merck
All Photos(1)

Key Documents

M1169

Sigma-Aldrich

7-Methoxycoumarin-4-acetyl-Asp-Glu-Val-Asp-Ala-Pro-(2,4-dinitrophenyl)Lys

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H62N10O22
CAS Number:
Molecular Weight:
1155.08
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available. Contact Technical Service

storage temp.

−20°C

SMILES string

COc1ccc2C(CC(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](C)C(=O)N3CCC[C@H]3C(=O)N[C@@H](CCCCNc4ccc(cc4[N+]([O-])=O)[N+]([O-])=O)C(O)=O)=CC(=O)Oc2c1

InChI

1S/C50H62N10O22/c1-24(2)43(57-44(69)31(14-15-39(62)63)54-46(71)33(22-40(64)65)53-38(61)18-26-19-42(68)82-37-21-28(81-4)11-12-29(26)37)48(73)56-34(23-41(66)67)45(70)52-25(3)49(74)58-17-7-9-35(58)47(72)55-32(50(75)76)8-5-6-16-51-30-13-10-27(59(77)78)20-36(30)60(79)80/h10-13,19-21,24-25,31-35,43,51H,5-9,14-18,22-23H2,1-4H3,(H,52,70)(H,53,61)(H,54,71)(H,55,72)(H,56,73)(H,57,69)(H,62,63)(H,64,65)(H,66,67)(H,75,76)/t25-,31-,32-,33-,34-,35-,43-/m0/s1

InChI key

YOUVSUDOGAVRQX-AEAFXXELSA-N

Amino Acid Sequence

MCA-Asp-Glu-Val-Asp-Ala-Pro-Lys-DNP

Biochem/physiol Actions

Fluorogenic substrate for caspase 3 and caspase 3-like enzymes. Cleavage at the P1 Asp residue results in continuous fluorescence at 392 nm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Enari et al.
Nature, 380(6576), 723-726 (1996-04-25)
Binding of Fas ligand or an agonistic anti-Fas antibody induces apoptosis in Fas-bearing cells. The interleukin-1Beta-converting enzyme (ICE) is a cysteine protease that is involved in apoptosis induced by various stimuli, including Fas-mediated apoptosis. Several ICE homologues have been identified

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service