Skip to Content
Merck
All Photos(1)

Key Documents

L7901

Sigma-Aldrich

Monoclonal Anti-LEF-1 (HMG DNA Binding Domain) antibody produced in mouse

~1 mg/mL, clone REMB6, purified immunoglobulin

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
₩1,298,693

₩1,298,693


Estimated to ship onJune 19, 2025Details



Select a Size

Change View
250 μG
₩1,298,693

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

₩1,298,693


Estimated to ship onJune 19, 2025Details


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

REMB6, monoclonal

mol wt

antigen 55-57 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1-5 μg/mL using whole extracts of human Jurkat cells or transiently expressed human LEF-1 in whole cell lysates from Cos-1 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... LEF1(51176)

Immunogen

full length recombinant LEF-1.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.5, containing 0.08% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marloes L de Groote et al.
International journal of cancer, 134(2), 280-290 (2013-07-09)
Ovarian cancer is a difficult-to-treat cancer with a 5-year survival rate of only ∼45%, due to late diagnosis and therapy resistance. In need of new therapeutic approaches, induction of intercellular adhesion molecule (ICAM)-1 expression might be of interest, since the
Elisabetta Lambertini et al.
BMC molecular biology, 11, 13-13 (2010-02-05)
Lymphoid Enhancer Factor-1 (Lef-1) is a member of a transcription factor family that acts as downstream mediator of the Wnt/beta-catenin signalling pathway which plays a critical role in osteoblast proliferation and differentiation. In a search for Lef-1 responsive genes in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service