Skip to Content
Merck
All Photos(2)

Key Documents

L7777

Sigma-Aldrich

Lacto-N-fucopentaose III

from human milk, ≥95% (HPAE)

Synonym(s):

β-D-Gal-(1→4)-[α-L-Fuc-(1→3)]-β-D-GlcNAc-(1→3)-β-D-Gal-(1→4)-D-Glc, LNFP III, Lex-lactose, Lewis-X pentasaccharide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H55NO25
CAS Number:
Molecular Weight:
853.77
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

biological source

human milk

Assay

≥95% (HPAE)

storage temp.

2-8°C

SMILES string

CC1OC(OC2C(NC(C)=O)C(OC(CO)C2OC3OC(CO)C(O)C(O)C3O)OC4C(O)C(CO)OC(OC(C(O)CO)C(O)C(O)C=O)C4O)C(O)C(O)C1O

InChI

1S/C32H55NO25/c1-8-16(42)20(46)22(48)30(51-8)57-27-15(33-9(2)39)29(54-14(7-38)26(27)56-31-23(49)21(47)18(44)12(5-36)52-31)58-28-19(45)13(6-37)53-32(24(28)50)55-25(11(41)4-35)17(43)10(40)3-34/h3,8,10-32,35-38,40-50H,4-7H2,1-2H3,(H,33,39)

InChI key

WMYQZGAEYLPOSX-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Md Ziaur Rahman et al.
Journal of biochemistry, 164(1), 53-63 (2018-02-15)
In a previous study, we molecular-characterized a tomato (Solanum lycopersicum) α1, 3/4-fucosidase (α-Fuc'ase Sl-1) encoded in a tomato gene (Solyc03g006980), indicating that α-Fuc'ase Sl-1 is involved in the turnover of Lea epitope-containing N-glycans. In this study, we have characterized another
Milady R Ninonuevo et al.
Journal of agricultural and food chemistry, 54(20), 7471-7480 (2006-09-28)
Oligosaccharides in human milk represent a group of bioactive molecules that have evolved to be an abundant and diverse component of human milk, even though they have no direct nutritive value to the infant. A recent hypothesis proposes that they
R Erney et al.
Advances in experimental medicine and biology, 501, 285-297 (2002-01-15)
Human milk is a unique reservoir of oligosaccharides. The presence of many of these oligosaccharides is determined genetically and is related to the Lewis blood group and secretor antigen status of each donor. A method to quantitate neutral human milk

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service