Skip to Content
Merck
All Photos(1)

Key Documents

L6654

Sigma-Aldrich

Lacto-N-hexaose

from human milk, ≥95%

Synonym(s):

β-D-Gal-(1→3)-β-D-GlcNAc-(1→3)-(β-D-Gal-[1→4]-β-D-GlcNAc-[1→6])-β-D-Gal-(1→4)-D-Glc, LNH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H68N2O31
CAS Number:
Molecular Weight:
1072.96
MDL number:
UNSPSC Code:
12352201

biological source

human milk

Assay

≥95%

storage temp.

2-8°C

SMILES string

CC(=O)NC1C(O)C(OC2OC(CO)C(O)C(O)C2O)C(CO)OC1OCC3OC(OC(C(O)CO)C(O)C(O)C=O)C(O)C(OC4OC(CO)C(O)C(OC5OC(CO)C(O)C(O)C5O)C4NC(C)=O)C3O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V K Dua et al.
Journal of chromatography, 328, 259-269 (1985-06-28)
The branched hexasaccharide whose trivial name is lacto-N-hexaose (LNH) as well as a mono- and a difuco-derivative were isolated from human breast milk of a Lewis a positive donor by high-performance liquid chromatography (HPLC). Reversed-phase HPLC on C18 columns which
C Dumon et al.
Glycoconjugate journal, 18(6), 465-474 (2002-06-27)
We report here the in vivo production of type 2 fucosylated-N-acetyllactosamine oligosaccharides in Escherichia coli. Lacto-N-neofucopentaose Galbeta1-4GlcNAcbeta1-3Galbeta1-4(Fucalpha1-3)Glc, lacto-N-neodifucohexaose Galbeta1-4(Fucalpha1-3)Glc-NAcbeta1-3Galbeta1-4(Fucalpha1-3)Glc, and lacto-N-neodifucooctaose Galbeta1-4GlcNAcbeta1-3Galbeta1-4(Fucalpha1-3)GlcNAcbeta1-3Galbeta1-4(Fucalpha1-3)Glc were produced from lactose added in the culture medium. Two of them carry the Lewis X human antigen.
Marion Rohmer et al.
Analytical chemistry, 82(9), 3719-3726 (2010-04-15)
Matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOF MS) is a widely used method in oligosaccharide analysis. Underivatized oligosaccharides are not well-suited for that purpose due to their low ionization efficiency; however, derivatization requires tedious sample purification steps which may lead

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service