Documents
HS120653
Slide Folder Clasp
assorted colors
About This Item
UNSPSC Code:
41121800
NACRES:
NB.16
material
assorted colors
packaging
pack of 100 ea
manufacturer/tradename
Heathrow Scientific 120653
General description
The Slide Folder Clasps provide an area for identification labels and secure the slide folder so that your slides do not fall out when the folder is transported.
Features and Benefits
  • Made from ABS
  • Ideal for histology, microbiology and pathology as well as multiple user research environments
  • Clasps can be labeled with a permanent marker or a label can be attached