HS120244
Microtube Rack
for tubes, 1.5 ‑ 2.0 mL, size 96 wells, white Nylon, pack of 4 racks
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
white Nylon
feature
for tubes 1.5 ‑ 2.0 mL
packaging
pack of 4 racks
size
96 wells
well diam.
11.1 mm
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- made from reinforced nylon
- superior chemical and heat resistance
- handles provide easy and safe transportation
- stackable two formats available for smaller and larger requirements
- temperature range -40 to 140C
- autoclavable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service