Key Documents
GE28-9042-71
illustra Plasmid Mini Spin
About This Item
UNSPSC Code:
41121800
NACRES:
NA.52
Pricing and availability is not currently available. Contact Technical Service
manufacturer/tradename
Cytiva
General description
illustra plasmidPrep Mini Spin Kit uses a simple plasmid DNA purification protocol involving a modified alkaline lysis procedure and a novel silica-based membrane to achieve highly efficient plasmid DNA purification. The typical plasmid DNA yield from a fresh 1.5 ml culture of E. coli containing a high copy number plasmid is approximately 9 μg. Instead of organic solvents, the kit contains chaotropic salts to denature protein components and promote selective binding of DNA to the novel silica membrane. The kit contains spin columns prepacked with a novel silica membrane, suspension, lysis, and neutralization buffers containing chaotropic salts, wash and elution buffers, microcentrifuge collection tubes, a full protocol booklet, and a detachable, quick reference protocol card.
Features and Benefits
Increased ease of use provided by minimal changes in pipetting volume from one part of the protocol to the next, fewer centrifugation steps, color-coded kit components, and lack of organic solvents.
Produces high-quality plasmid DNA with excellent reproducibility for use in cloning, restriction enzyme digestion, PCR amplification, and DNA sequencing.
Total preparation time of less than 10 min represents a 50% reduction in protocol time.
Legal Information
illustra is a trademark of Cytiva
Storage Class Code
8B - Non-combustible corrosive hazardous materials
Choose from one of the most recent versions:
Certificates of Analysis (COA)