Skip to Content
Merck
All Photos(1)

Documents

EP022491288

Eppendorf® ep Dualfilter T.I.P.S.®

PCR clean, sterile, volume 0.5-10 mL, L 243 mm, turquoise, colorless tip

Synonym(s):

filter tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601

material

colorless tip
polypropylene
turquoise

sterility

sterile

feature

DNase free
PCR clean
RNase free
endotoxin free

packaging

pack of 100 ea

manufacturer/tradename

Eppendorf® 022491288

L

243 mm

volume

0.5-10 mL

General description

ep Dualfilter T.I.P.S.®, PCR clean and sterile, 0.5 – 10 mL L, 243 mm, turquoise, colorless tips, 100 tips, individually packed
  • Two filter layers provide dual protection against aerosols1) and biomolecules to guard your pipette and sample
  • Free of PCR inhibiting additives provide for reliable downstream results
  • Available in PCR clean/Sterile (sterile and pyrogen-free)
  • Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality
  • Lot-specific certificates are available at (URL: https://www.eppendorf.com/certificates/)[certificates homepage]
  • Certificate of filter efficiency acc. to EN 1822 available (URL: https://www.eppendorf.com/fileadmin/Main/05-Files/INT/Service___Support/CoQ_EPA_HEPA_Standard_of_epDualfilter_T.I.P.S._March_2020.pdf)[here]

1) An aerosol is a colloid of fine solid particles or liquid droplets, in air or another gas.

Features and Benefits

  • Two filter layers provide dual protection against aerosols1) and biomolecules to guard your pipette and sample
  • Free of PCR inhibiting additives provide for reliable downstream results
  • Available in PCR clean/Sterile (sterile and pyrogen-free)
  • Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality

  • Two filter layers provide dual protection against aerosols1) and biomolecules to guard your pipette and sample
  • Free of PCR inhibiting additives provide for reliable downstream results
  • Available in PCR clean/Sterile (sterile and pyrogen-free)
  • Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service