EP0030108116
Eppendorf® Protein LoBind tubes
PCR clean, withstand, 18,000 x g, capacity 1.5 mL, pack of 100 ea
Synonym(s):
eppendorf tubes, microcentrifuge tubes, microfuge tubes
About This Item
Recommended Products
material
cap (plug seal)
colorless cap
colorless tube
conical bottom
polypropylene cap
polypropylene tube
sterility
non-sterile
feature
PCR clean
graduations
packaging
pack of 100 ea
manufacturer/tradename
Eppendorf® 0030108116
technique(s)
PCR: suitable
capacity
1.5 mL
binding type
low binding surface
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- Eppendorf® LoBind material ensures optimized sample recovery for improved assay results.
- Free of surface coating (e.g., silicone) to minimize the risk of sample interference.
- Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
- Available in tube formats for easy-up scaling.
- Precise lid sealing to minimize evaporation.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service