EP0030000897
Eppendorf® epT.I.P.S.® standard
volume range 2-300 μL, yellow polypropylene, pack of 1000 ea (2 bags × 500 tips)
Synonym(s):
Eppendorf pipette tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
yellow polypropylene
sterility
non-sterile
packaging
pack of 1000 ea (2 bags × 500 tips)
manufacturer/tradename
Eppendorf® 0030000897
availability
available only in (India)
maximum volume
300 μL
volume range
2-300 μL
Looking for similar products? Visit Product Comparison Guide
General description
Eppendorf Quality ensures efficient function with tight fitting and precision and precise shape. Low wetting and high chemical and thermal resistance. Good centrifugation stability and excellent material transparency. Color coded pack is dark green containing colorless tips.
Application
Liquid handling
Features and Benefits
- Tips are autoclave friendly at 121 °C in 20 min
- High-quality and original pipettes packed in resealable bags
- Easy to work with small sample volumes due to fine retracted or extended tips
- Original, high-quality Eppendorf pipette tips packaged in resealable bags
- Available in sizes from 10 µL to 10 mL in Eppendorf Quality purity grade epT.I.P.S. Standard 200 µL, 300 µL and 1,000 µL tips are also available color-coded yellow and blue
- epT.I.P.S. Standard pipette tips can be autoclaved at 121 °C in 20 min.
- Fine retracted or extended tips simplify work with small volumes
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service