D7669
Dimethyl isosorbide
~98% (GC)
Synonym(s):
Isosorbide dimethyl ether
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
EC Number:
UNSPSC Code:
12352200
Recommended Products
Assay
~98% (GC)
storage temp.
2-8°C
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service