Skip to Content
Merck
All Photos(1)

Documents

KHOS-312H

86112811, human bone, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

KHOS-312H, 86112811

biological source

human bone

growth mode

Adherent

karyotype

2n = 46

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human Caucasian osteosarcoma

Cell Line Description

A revertant line of the KHOS/NP cell line. Established after overnight incubation at 40.5°C and then sub-cloned at 36°C. The line has similar properties to the parent HOS cell line. Cells are non-tumourigenic in nude mice and no longer possess a rescuable MSV genome.

Application

Tumourigenicity, transformation, biochemical and virus studies: MSV

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 12
D13S317: 12
D16S539: 10,13
D5S818: 13
D7S820: 11,12
THO1: 6
TPOX: 8,11
vWA: 18

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service