Skip to Content
Merck
All Photos(1)

Documents

MRC-5 Cell Line human

84101801, human lung

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

product name

MRC-5 Cell Line human, 84101801, From fetal lung fibroblast-like

biological source

human lung

form

cell suspension

shipped in

dry ice

storage temp.

−196°C

Application

Established from normal lung tissue of a 14-week-old male fetus. The cells undergo 60-70 population doublings before senescence. The virus susceptibility of this line to human viruses is similar to that of WI-38 cells.
Subculture: Split at 70-80% confluency, approx. 1:3 to 1:6, seed at 2-4 × 104 viable cells/cm2. Trypsinize using 0.25% solution, with or without EDTA, 37 °C and 5% CO2.
Recommended culture medium: EMEM (EBSS) + 2 mM L-Glutamine + 1% NEAA (non-essential amino acids) + 10% FBS.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service