Skip to Content
Merck
All Photos(1)

Documents

A112030

Sigma-Aldrich

DMT-dA(tac) Phosphoramidite

configured for ABI

Synonym(s):

DMT-dA(tac) amidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H62N7O8P
CAS Number:
Molecular Weight:
944.06
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.51

type

for DNA synthesis

Quality Level

product line

Proligo Reagents

Assay

≥98% (31P-NMR)
≥98.0% (reversed phase HPLC)

form

powder

technique(s)

oligo synthesis: suitable

λ

conforms (UV/VIS Identity)

compatibility

configured for ABI

nucleoside profile

base: deoxyadenosine
base protecting group: TAC
2' protecting group: none
5' protecting group: DMT
deprotection: fast

storage temp.

-10 to -25°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4c(NC(=O)COc5ccccc5)ncnc34)(c6ccccc6)c7ccc(OC)cc7

InChI

1S/C48H54N7O8P/c1-33(2)55(34(3)4)64(61-27-13-26-49)63-41-28-44(54-32-52-45-46(50-31-51-47(45)54)53-43(56)30-59-40-16-11-8-12-17-40)62-42(41)29-60-48(35-14-9-7-10-15-35,36-18-22-38(57-5)23-19-36)37-20-24-39(58-6)25-21-37/h7-12,14-25,31-34,41-42,44H,13,27-30H2,1-6H3,(H,50,51,53,56)/t41-,42+,44+,64?/m0/s1

InChI key

INUFZOANRLMUCW-SDNJGBRXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Proligo′s DNA phosphoramidites lead to high-yield and high-quality oligonucleotides due to their high coupling efficiency. The deprotection step of automated oligonucleotide synthesis is integral to synthesis time and final product quality. The high coupling efficiency of Proligo′s DNA phosphoramidites leads to high-yield and high-quality oligonucleotides. Substitution of standard protecting groups with the labile TAC (tert.-butylphenoxyacetyl) protecting group results in ultra-fast and easy deprotection under mild conditions, suitable for oligonucleotides with base labile monomers and reporters as well as in-situ synthesis schemes on glass surfaces. Key Features of TAC Chemistry:
  • Deprotection of the TAC group is ultra-fast: complete deprotection in concentrated ammonia occurs within 15 minutes at 55 °C or two hours atroom temperature
  • Compatible with the AMA deprotection reagent (a mixture of ≥25%ammonia in water with 40% aqueous methylamine I/I, v/v)
  • Highly soluble in acetonitrile. No need to add co-solvents such as dimethylformamide or methylene chloride
  • Suitable for the synthesis of oligomers with base-labile units e.g., dyes and modifiers, because of less exposure to ammonia and the possibility ofroom temperature deprotection
  • No change is required in the reagents commonly used for DNA synthesis,except that Proligo′s Fast Deprotection Cap A solution is used instead ofCap A solution
  • The application of dA(tac) minimizes depurination and improves thequality of oligonucleotides
DMT-dA(tac) Phosphoramidite configured for ABI Synthesizers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service