storage temp.
−20°C
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Michael J Storek et al.
Organic letters, 4(22), 3867-3869 (2003-02-26)
[formula: see text] 5-Amino-2'-deoxyuridine 5'-triphosphate, an analogue of deoxythymidine triphosphate, was synthesized and found to be a substrate of Taq DNA polymerase. The DNA-borne analogue underwent selective chemical reaction with permanganate. The use of 5-amino-dU as an interference probe was
V S Rana et al.
Organic letters, 1(4), 631-633 (2000-05-24)
[formula: see text] 5-Amino-dU, a designed nucleoside analogue, when placed in the central strand of DNA triple helix, recognizes all four bases A, G, C, and T in the third strand, with a selectivity based on the orientation (parallel/antiparallel) of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service