Skip to Content
Merck
All Photos(1)

Key Documents

43605

Sigma-Aldrich

1,1′-Dipropyl-3,3,3′,3′-tetramethylindocarbocyanine iodide

suitable for fluorescence, ≥98.0% (TLC)

Synonym(s):

DiIC3(3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H37IN2
CAS Number:
Molecular Weight:
540.52
Beilstein:
9604018
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98.0% (TLC)

solubility

DMSO: soluble
methanol: soluble

fluorescence

λex 548 nm; λem 599 nm in phosphate buffer/SDS pH 7.0

suitability

suitable for fluorescence

SMILES string

[I-].CCCN1c2ccccc2C(C)(C)\C1=C\C=C\C3=[N+](CCC)c4ccccc4C3(C)C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service