Skip to Content
Merck
All Photos(1)

Key Documents

42726

Supelco

3-(4,6-Difluorotriazinylamino)-7-methoxycoumarin

≥95% (coupling to amines)

Synonym(s):

FAMC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8F2N4O3
CAS Number:
Molecular Weight:
306.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (coupling to amines)

fluorescence

λex 340 nm; λem 421 nm in 0.1 M phosphate pH 7.0

SMILES string

COc1ccc2C=C(Nc3nc(F)nc(F)n3)C(=O)Oc2c1

InChI

1S/C13H8F2N4O3/c1-21-7-3-2-6-4-8(10(20)22-9(6)5-7)16-13-18-11(14)17-12(15)19-13/h2-5H,1H3,(H,16,17,18,19)

InChI key

HHJOBSJNFNVLNR-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service