Skip to Content
Merck
All Photos(3)

Key Documents

VT000104

Millipore

Enterococcus faecium WDCM 00010 Vitroids

130-300 CFU mean value range, certified reference material, suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

form

disc (hydrosoluble)

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

storage temp.

−20°C

suitability

Enterococcus spp.

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Download your certificate at: http://www.sigma-aldrich.com.

Please specify if you require a single batch - in the remark field when ordering (only for orders with more than 15 packs)
Vitroids discs contain Enterococcus faecalis WDCM 000010 viable microorganisms (130-300 CFU) in a certified quantity (generally accredited according to ISO/IEC 17025), produced under reproducible conditions compliant with ISO 17034:2016 using authenticated strains from CECT®.
The discs consist of pure cultures of bacteria in a solid matrix, stable for at least one year and are in a viable state with a shelf life of 1-3 years.
Each batch is provided with a comprehensive certificate of analysis that specifies the mean number of colony-forming units (CFU), an expanded uncertainty about the mean, details about the method used to determine the product data and the number of passages (subcultures) from the original strain.
Vitroids are derived from a traceable culture obtained freeze-dried from CECT®.

Application

  • Daily quality control
  • Performance testing of media acc. ISO 11133
  • Validation of new methods
  • Materials for proficiency testing or ring trials
  • Method development
  • Staff training
  • Starter cultures

Features and Benefits

  • Defined CFU range and low standard deviation
  • Fast, reliable and easy to use

Other Notes

ATCC® 19434, NCTC 7171, NCPF -, DSM 20477, LMG 11423, IHEM -, CIP 103014T
Origin Strain No. CECT® 410

Legal Information

Derived from CECT® strains
ATCC is a registered trademark of American Type Culture Collection
CECT is a registered trademark of Universitat de Valencia
Vitroids is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service