Skip to Content
Merck
All Photos(1)

Key Documents

PHR8641

Supelco

Testosterone acetate

certified reference material, pharmaceutical secondary standard

Synonym(s):

17β-Acetoxy-4-androsten-3-one, 17β-Hydroxy-4-androsten-3-one 17-acetate, 4-Androsten-17β-ol-3-one 17-acetate, Deposteron

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
₩1,248,653

₩1,248,653


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
₩1,248,653

About This Item

Empirical Formula (Hill Notation):
C21H30O3
CAS Number:
Molecular Weight:
330.46
Beilstein:
2062555
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

₩1,248,653


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

API family

testosterone

packaging

pkg of 100MG

application(s)

pharmaceutical

storage temp.

-10 to -25°C

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@H](CC[C@@]24[H])OC(C)=O

InChI key

DJPZSBANTAQNFN-PXQJOHHUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service