Skip to Content
Merck
All Photos(1)

Documents

92081

Supelco

Baicalein

analytical standard

Synonym(s):

5,6,7-Trihydroxyflavone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
Beilstein:
272683
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

256-271 °C (lit.)

format

neat

SMILES string

Oc1cc2OC(=CC(=O)c2c(O)c1O)c3ccccc3

InChI

1S/C15H10O5/c16-9-6-11(8-4-2-1-3-5-8)20-12-7-10(17)14(18)15(19)13(9)12/h1-7,17-19H

InChI key

FXNFHKRTJBSTCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Gao et al.
ACS chemical neuroscience, 9(7), 1714-1724 (2018-04-19)
Baicalein, a flavonoid derived from the roots of Scutellariae baicalensis Georgi, has shown health benefits for an array of human diseases including dementia. The senescence-accelerated mouse prone 8 (SAMP8) strain is extensively used as a senile dementia model. To further
Andreas Körner et al.
Nature communications, 10(1), 633-633 (2019-02-09)
The bidirectional communication between the immune and nervous system is important in regulating immune responses. Here we show that the adrenergic nerves of sympathetic nervous system orchestrate inflammation resolution and regenerative programs by modulating repulsive guidance molecule A (RGM-A). In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service