Skip to Content
Merck
All Photos(1)

Documents

91732

Sigma-Aldrich

Sodium trifluoroacetate

BioUltra, ≥99.0% (NT)

Synonym(s):

Trifluoroacetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COONa
CAS Number:
Molecular Weight:
136.01
Beilstein:
3597949
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

product line

BioUltra

Assay

≥99.0% (NT)

impurities

insoluble matter, passes filter test

mp

205-207 °C (dec.) (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

density

1.49 g/mL (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤20 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.055
λ: 280 nm Amax: ≤0.035

SMILES string

[Na+].[O-]C(=O)C(F)(F)F

InChI

1S/C2HF3O2.Na/c3-2(4,5)1(6)7;/h(H,6,7);/q;+1/p-1

InChI key

UYCAUPASBSROMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Competitive inhibitor of fluoroacetate fluorohydrolase from pseudomonad species

replaced by

Product No.
Description
Pricing

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

THE ENZYMATIC CLEAVAGE OF THE CARBON-FLUORINE BOND IN FLUOROACETATE.
P GOLDMAN
The Journal of biological chemistry, 240, 3434-3438 (1965-08-01)
Maxime Mourer et al.
Bioorganic & medicinal chemistry, 17(15), 5496-5509 (2009-07-15)
The tetra-para-guanidinoethyl-calix[4]arene, its distally-disubstituted ether derivatives involving 2,2'-bithiazolyl- or 2,2'-bipyridyl-methyl groups, as well as the para-guanidinoethylphenol and its analogous derivatives have been synthesized, fully characterized and evaluated as antibacterial agents towards both gram positive and gram negative reference bacteria. The
Hugues Massimba Dibama et al.
Bioorganic & medicinal chemistry letters, 19(10), 2679-2682 (2009-04-28)
A water-soluble calixarene-based heterocyclic podand incorporating a quinolone antibiotic subunit, the nalidixic acid, was synthesised and fully characterised. Its prodrug behaviour was assessed in vitro by HPLC, demonstrating the release of the tethered quinolone in model biological conditions. Microbiological studies
Cristina Japu et al.
Biomacromolecules, 16(3), 868-879 (2015-01-27)
Biotechnologically accessible 1,4-butanediol and vegetal oil-based diethyl sebacate were copolymerized with bicyclic acetalized D-glucose derivatives (Glux) by polycondensation both in the melt at high temperature and in solution at mild temperature mediated by polymer-supported Candida antarctica lipase B (CALB). Two
Inara de Aguiar et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 70, 45-54 (2015-02-02)
Despite the resistance developed by the Mycobacterium tuberculosis (MTb) strains, isoniazid (INH) has been recognized as one of the best drug for treatment of Tuberculosis (Tb). The coordination of INH to ruthenium metal centers was investigated as a strategy to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service