Skip to Content
Merck
All Photos(1)

Key Documents

91674

Sigma-Aldrich

1,1,1-Trifluoro-2,4-pentanedione

purum, ≥98.0% (GC)

Synonym(s):

α,α,α-Trifluoroacetylacetone, 1,1,1-Trifluoroacetylacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COCF3
CAS Number:
Molecular Weight:
154.09
Beilstein:
1705177
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (GC)

color

deep brown-yellow

refractive index

n20/D 1.388 (lit.)
n20/D 1.389

bp

105-107 °C (lit.)

density

1.27 g/mL at 25 °C (lit.)

SMILES string

CC(=O)CC(=O)C(F)(F)F

InChI

1S/C5H5F3O2/c1-3(9)2-4(10)5(6,7)8/h2H2,1H3

InChI key

SHXHPUAKLCCLDV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service