Skip to Content
Merck
All Photos(1)

Key Documents

74878

Sigma-Aldrich

N-Octanoyl-β-D-glucosylamine

≥98.0% (TLC)

Synonym(s):

N-(β-D-Glucopyranosyl)octanamide, NOGA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H27NO6
CAS Number:
Molecular Weight:
305.37
Beilstein:
4321579
UNSPSC Code:
12161900

description

non-ionic

Assay

≥98.0% (TLC)

optical activity

[α]20/D +18±1°, c = 1% in DMF

storage temp.

2-8°C

SMILES string

CCCCCCCC(=O)N[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service