Skip to Content
Merck
All Photos(1)

Key Documents

46045

Sigma-Aldrich

Ethynylmagnesium bromide solution

technical, ~0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CMgBr
CAS Number:
Molecular Weight:
129.24
Beilstein:
3600874
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

grade

technical

concentration

~0.5 M in THF

density

0.94 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

Br[Mg]C#C

InChI

1S/C2H.BrH.Mg/c1-2;;/h1H;1H;/q;;+1/p-1

InChI key

HUGJUYPSXULVQQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Caution

can precipitate on storage

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service