Skip to Content
Merck
All Photos(1)

Key Documents

43924

Supelco

3-[(Dimethylamino)methyl]-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one hydrochloride

pharmaceutical impurity standard

Synonym(s):

Ondansetron Related Compound A (USP), Ondansetron impurity A (PhEur), 3-[(Dimethylamino)methyl]-1,2,3,9-tetrahydro-9-methyl-4H-carbazol-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20N2O · HCl
CAS Number:
Molecular Weight:
292.80
Beilstein:
11612579
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% HPLC impurities
≤5% solvents
≤5% water

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CN(C)CC1CCc2c(C1=O)c3ccccc3n2C

InChI

1S/C16H20N2O.ClH/c1-17(2)10-11-8-9-14-15(16(11)19)12-6-4-5-7-13(12)18(14)3;/h4-7,11H,8-10H2,1-3H3;1H

InChI key

GFQLILQFQHMEPF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Met. Corr. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service