Key Documents
26163
Hamilton® plunger assemblies with tips for series 1000, 1700, and 1800
for use with Hamilton 1010-LT-LTN-RN-TLL, pkg of 1 ea
About This Item
UNSPSC Code:
41122001
material
PTFE plunger tip
size
10 mL
description
1010 LT-LTN-RN-TLL
feature
Includes plunger tip, plunger and O-ring (if required).
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 13231
compatibility
for use with Hamilton 1010-LT-LTN-RN-TLL
General description
For field repair of Hamilton Series 1000, 1700 and 1800 syringes.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)