Skip to Content
Merck
All Photos(1)

Key Documents

25220

Sigma-Aldrich

4-Chloro-1-methylpiperidine hydrochloride

technical, ≥97.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12ClN · HCl
CAS Number:
Molecular Weight:
170.08
Beilstein:
3677071
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥97.0% (AT)

mp

158-162 °C (lit.)
158-162 °C

SMILES string

Cl[H].CN1CCC(Cl)CC1

InChI

1S/C6H12ClN.ClH/c1-8-4-2-6(7)3-5-8;/h6H,2-5H2,1H3;1H

InChI key

QDOJNWFOCOWAPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service