Skip to Content
Merck
All Photos(1)

Key Documents

24190

Sigma-Aldrich

Chlorocyclopentane

purum, ≥99.0% (GC)

Synonym(s):

Cyclopentyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9Cl
CAS Number:
Molecular Weight:
104.58
Beilstein:
1900557
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥99.0% (GC)

refractive index

n20/D 1.4512 (lit.)
n20/D 1.452

bp

114 °C (lit.)

density

1.005 g/mL at 25 °C (lit.)

SMILES string

ClC1CCCC1

InChI

1S/C5H9Cl/c6-5-3-1-2-4-5/h5H,1-4H2

InChI key

NDTCXABJQNJPCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service