21748-U
Hamilton® needles for LT and TLL syringes
Metal Hub (N), needle size 22s ga (bevel tip), needle L 51 mm (2 in.), pkg of 6 ea
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
material
bevel tip stainless steel needle (point style 2)
needle size
22s ga (bevel tip)
feature
Metal Hub (N)
packaging
pkg of 6 ea
manufacturer/tradename
Hamilton 90038
needle L
51 mm (2 in.)
compatibility
for use with Hamilton LT and TLL
Looking for similar products? Visit Product Comparison Guide
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service