Skip to Content
Merck
All Photos(1)

Documents

20050

Sigma-Aldrich

2-Iodo-2-methylpropane

technical, ≥90% (GC)

Synonym(s):

tert-Butyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3Cl
CAS Number:
Molecular Weight:
184.02
Beilstein:
505950
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥90% (GC)

contains

silver wool as stabilizer

refractive index

n20/D 1.491 (lit.)
n20/D 1.491

bp

99-100 °C (lit.)

mp

−38 °C (lit.)

density

1.544 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)I

InChI

1S/C4H9I/c1-4(2,3)5/h1-3H3

InChI key

ANGGPYSFTXVERY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Iodo-2-methylpropane was degraded by reductive dehalogenation with nickel-aluminum alloy in potassium hydroxide solution.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Lunn et al.
American Industrial Hygiene Association journal, 52(6), 252-257 (1991-06-01)
Two techniques were investigated for degrading a number of halogenated compounds of commercial and research importance. Reductive dehalogenation with nickel-aluminum alloy in potassium hydroxide solution was used to degrade iodomethane, chloroacetic acid, trichloroacetic acid, 2-chloroethanol, 2-bromoethanol, 2-chloroethylamine, 2-bromoethylamine, 1-bromobutane, 1-iodobutane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service