17550
2-(Bromomethyl)acrylic acid
purum, ≥97.0% (T)
Synonym(s):
α-(Bromomethyl)acrylic acid, 2-(Bromomethyl)propenoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH2=C(CH2Br)COOH
CAS Number:
Molecular Weight:
164.99
Beilstein:
2426204
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
grade
purum
Assay
≥97.0% (T)
mp
68-73 °C
70-73 °C (lit.)
storage temp.
2-8°C
SMILES string
OC(=O)C(=C)CBr
InChI
1S/C4H5BrO2/c1-3(2-5)4(6)7/h1-2H2,(H,6,7)
InChI key
NOOYFQLPKUQDNE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
Preparation of the corresponding Wittig-Horner reagents; The ethyl ester is used for Reformatskii-type reactions giving α-methylene-γ-lactones
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
H. Duttmann et al.
Chemische Berichte, 112, 3480-3480 (1979)
K. Ramarajan et al.
Organic Syntheses, 59, 2080-2080 (1980)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service