14423
Acetaldehyde, DAIH derivative
analytical standard
Synonym(s):
2-Diphenylacetyl-3-(ethylidene-hydrazono)indan-1-one, 2-Diphenylacetyl-indan-1,3-dione-1-ethylidenehydrazone
About This Item
Recommended Products
grade
analytical standard
Assay
≥98.0% (HPLC)
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
neat
SMILES string
C\C=N\N=C1/C(C(=O)C(c2ccccc2)c3ccccc3)C(=O)c4ccccc14
InChI
1S/C25H20N2O2/c1-2-26-27-23-19-15-9-10-16-20(19)24(28)22(23)25(29)21(17-11-5-3-6-12-17)18-13-7-4-8-14-18/h2-16,21-22H,1H3/b26-2+,27-23-
InChI key
IKWSJQIXRGFICA-JPUYTKAPSA-N
Application
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service