Skip to Content
Merck
All Photos(1)

Key Documents

10885

Sigma-Aldrich

2-Methylene-1,3-dioxepane

produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0% (GC)

Synonym(s):

Ketene tetramethylene acetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
MDL number:
UNSPSC Code:
12352103

grade

produced by Wacker Chemie AG, Burghausen, Germany

Assay

≥98.0% (GC)

bp

62-64 °C/50 hPa

storage temp.

2-8°C

SMILES string

C=C1OCCCCO1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service